Assay Details
Target Gene Details
Entrez Gene ID: | 217558 |
Gene Name: | G2/M-phase specific E3 ubiquitin ligase |
Gene Aliases: |
6030408C04Rik, 9030416F18, AW046379, D930034K21Rik, mKIAA1333 |
Location: |
Chr.12:51348230-51376986 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
G2e3 | NM_001015099.2 | NP_001015099.2 | ||
NM_001167963.1 | NP_001161435.1 | |||
NM_001167964.1 | NP_001161436.1 | |||
AK031333.1 | ||||
AK033513.1 | ||||
AK084256.1 | BAC39150.1 | |||
AK129332.1 | BAC98142.1 | |||
BC086455.1 | AAH86455.1 |
More Information
Set Membership: |
Intragenic Non-exonic |