Assay Details
Target Gene Details
Entrez Gene ID: | 15251 |
Gene Name: | hypoxia inducible factor 1, alpha subunit |
Gene Aliases: |
AA959795, HIF1alpha, MOP1, bHLHe78 |
Location: |
Chr.12:73901329-73947530 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hif1a | NM_010431.2 | 2 | 557 | NP_034561.2 |
AF003695.1 | 2 | 391 | AAC53455.1 | |
AK017853.1 | 2 | 418 | BAB30975.1 | |
AK034087.1 | 2 | 523 | BAC28578.1 | |
AK138944.1 | 2 | 424 | BAE23829.1 | |
AK150367.1 | 2 | 424 | BAE29500.1 | |
BC026139.1 | 2 | 409 | AAH26139.1 | |
BY725437.1 | 2 | 557 | ||
X95002.1 | 1 | 71 | CAA64458.1 | |
X95580.1 | 2 | 223 | CAA64833.1 |
More Information
Set Membership: |
Intragenic Exonic |