Assay Details
Target Gene Details
Entrez Gene ID: | 67216 |
Gene Name: | membrane bound O-acyltransferase domain containing 2 |
Gene Aliases: |
2810049G06Rik, AU022889, AW547221, LPCAT4, Moact2, Oact2 |
Location: |
Chr.12:24831487-24964400 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mboat2 | NM_001083341.1 | 7 | 748 | NP_001076810.1 |
NM_026037.3 | 8 | 844 | NP_080313.2 | |
AB297383.1 | 8 | 743 | ||
AB305046.1 | 8 | 743 | ||
AK012931.1 | 7 | 732 | BAB28556.1 | |
AK031824.1 | 7 | 733 | BAC27567.1 | |
AK033106.1 | 5 | 508 | BAC28154.1 | |
AK034873.1 | 5 | 532 | BAC28863.1 | |
AK076045.1 | 7 | 738 | BAC36144.1 | |
BC025020.1 | 7 | 698 | AAH25020.1 |
More Information
Set Membership: |
Intragenic Exonic |