Assay Details
Target Gene Details
Entrez Gene ID: | 72258 |
Gene Name: | potassium channel, subfamily K, member 10 |
Gene Aliases: |
1700024D23Rik, 3010005K24Rik, Trek2 |
Location: |
Chr.12:98429432-98577940 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnk10 | NM_029911.4 | 1 | 84 | NP_084187.2 |
AK036066.1 | 1 | 358 | BAC29295.1 | |
AK082153.1 | 1 | 85 | BAC38424.1 | |
BC132487.1 | ||||
BC137869.1 |
More Information
Set Membership: |
Intragenic Exonic |