Assay Details
Target Gene Details
Entrez Gene ID: | 211151 |
Gene Name: | churchill domain containing 1 |
Gene Aliases: |
1700030P15Rik, My015, chch |
Location: |
Chr.12:76765573-76783180 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Churc1 | NM_206534.1 | NP_996257.1 | ||
AK085076.1 | 1 | 86 | ||
BC019472.1 | AAH19472.1 | |||
BC076499.1 | AAH76499.1 |