Assay Details
Target Gene Details
Entrez Gene ID: | 21408 |
Gene Name: | zinc finger protein 354A |
Gene Aliases: |
AW488485, Tcf17, Znf354a, kid1 |
Location: |
Chr.11:51059244-51072799 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp354a | NM_009329.3 | NP_033355.2 | ||
AB221586.1 | ||||
AF184111.1 | AAF01033.1 | |||
AK030469.1 | ||||
BC050843.1 | AAH50843.1 | |||
BC087540.1 | AAH87540.1 | |||
L77247.1 | AAA96309.1 |