Assay Details
Target Gene Details
Entrez Gene ID: | 403205 |
Gene Name: | anterior gradient 3 |
Gene Aliases: |
BCMP11, E030025L21Rik, Gm888 |
Location: |
Chr.12:35925621-35949737 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agr3 | NM_207531.3 | 8 | 656 | NP_997414.2 |
AK053177.1 | 8 | 585 | BAC35297.1 | |
AW490945.1 | 1 | 187 | ||
BC023499.1 | 8 | 657 | AAH23499.1 |
More Information
Set Membership: |
Intragenic Exonic |