Assay Details
Target Gene Details
Entrez Gene ID: | 23795 |
Gene Name: | anterior gradient 2 |
Gene Aliases: |
Agr2h, Gob-4, HAG-2, XAG-2, mAG-2 |
Location: |
Chr.12:35992925-36004081 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agr2 | NM_011783.2 | NP_035913.1 | ||
AB016592.1 | BAA32044.1 | |||
AF044262.1 | AAC72705.1 | |||
AK007677.1 | BAB25181.1 | |||
AK008296.1 | ||||
BC013334.1 | AAH13334.1 |
More Information
Set Membership: |
Intragenic Non-exonic |