Assay Details
Target Gene Details
Entrez Gene ID: | 66272 |
Gene Name: | cytochrome c oxidase assembly protein 16 |
Gene Aliases: |
1810020G14Rik, 1810055I05Rik, BB388670 |
Location: |
Chr.12:81470704-81485110 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cox16 | NM_025461.5 | 1 | 101 | NP_079737.1 |
AK007450.1 | 1 | 78 | BAB25045.1 | |
AK007562.1 | 1 | 90 | BAB25110.1 | |
AK008486.1 | 1 | 66 | BAB25693.1 | |
AK040003.1 | 1 | 103 | BAC30495.1 | |
AK054491.1 | 1 | 77 | ||
AK082374.1 | 1 | 102 | BAC38480.1 | |
AK084315.1 | 1 | 98 |