Assay Details
Target Gene Details
Entrez Gene ID: | 21975 |
Gene Name: | topoisomerase (DNA) III alpha |
Gene Aliases: |
- |
Location: |
Chr.11:60739473-60777365 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Top3a | NM_009410.2 | 7 | 1042 | NP_033436.1 |
AB006074.1 | 7 | 1043 | BAA25662.1 | |
AK029987.1 | 4 | 435 | BAC26718.1 | |
AK037640.1 | 7 | 943 | BAE20515.1 | |
AK164443.1 | 7 | 943 | BAE37792.1 | |
AK170291.1 | 8 | 849 | BAE41691.1 | |
AK171286.1 | 8 | 849 | BAE42369.1 | |
BC099689.1 | 7 | 857 | AAH99689.1 |
More Information
Set Membership: |
Intragenic Exonic |