Assay Details
Target Gene Details
Entrez Gene ID: | 70373 |
Gene Name: | G patch domain containing 2 like |
Gene Aliases: |
1700020O03Rik, 4732487G21Rik, A630090I05Rik, Kiaa1152 |
Location: |
Chr.12:86241838-86291793 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpatch2l | NM_027405.2 | NP_081681.2 | ||
AK006185.1 | BAB24446.1 | |||
AK031712.1 | BAC27527.1 | |||
AK129299.1 | BAC98109.1 | |||
AK147916.1 | BAE28226.1 | |||
AK181673.1 | ||||
AK203506.1 | ||||
BC048169.1 | AAH48169.1 | |||
BC058256.1 | AAH58256.1 |
More Information
Set Membership: |
Intragenic Non-exonic |