Assay Details
Target Gene Details
Entrez Gene ID: | 11491 |
Gene Name: | a disintegrin and metallopeptidase domain 17 |
Gene Aliases: |
CD156b, Tace |
Location: |
Chr.12:21323509-21373632 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adam17 | NM_001277266.1 | 14 | 1858 | NP_001264195.1 |
NM_001291871.1 | 13 | 1820 | NP_001278800.1 | |
NM_009615.6 | 13 | 1801 | NP_033745.4 | |
NR_102380.1 | 10 | 1444 | ||
NR_120376.1 | 13 | 1801 | ||
AB021709.1 | 13 | 1762 | BAA78578.1 | |
AJ007365.1 | 13 | 1601 | CAA07480.1 | |
AK137183.1 | 6 | 2010 | ||
AK137693.1 | 13 | 1708 | BAE23464.1 | |
AK139471.1 | 13 | 1707 | BAE24023.1 | |
AK142441.1 | 13 | 1760 | ||
AK144161.1 | 14 | 1788 | BAE25740.1 | |
AK149288.1 | 13 | 1704 | BAE28792.1 | |
AK165739.1 | BAE38362.1 | |||
AK211820.1 | 2 | 87 | ||
BC094655.1 | 13 | 1715 | AAH94655.1 | |
BC114999.1 | 13 | 1661 | AAI15000.1 | |
BC138416.1 | 13 | 1680 | ||
BC138420.1 | 13 | 1680 | ||
BC145270.1 | 13 | 1680 | ||
U69613.1 | 13 | 1669 | AAD09627.1 | |
U69614.1 | 13 | 1753 | AAD09628.1 |
More Information
Set Membership: |
Intragenic Exonic |