Assay Details
Target Gene Details
Entrez Gene ID: | 20913 |
Gene Name: | syntaxin binding protein 4 |
Gene Aliases: |
6030470M02Rik, Synip |
Location: |
Chr.11:90476492-90638123 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stxbp4 | NM_011505.2 | 3 | 212 | NP_035635.1 |
AF152924.1 | 3 | 202 | AAD43533.1 | |
AK012293.1 | 3 | 170 | ||
AK049001.1 | 3 | 181 | ||
BC032881.1 | 3 | 166 | AAH32881.1 | |
BY720472.1 | 3 | 213 |