Assay Details
Target Gene Details
Entrez Gene ID: | 18293 |
Gene Name: | oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide) |
Gene Aliases: |
2210403E04Rik, 2210412K19Rik, AA409584, d1401, mKIAA4192 |
Location: |
Chr.11:6291593-6359094 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ogdh | NM_001252282.1 | 22 | 2884 | NP_001239211.1 |
NM_001252283.1 | 22 | 2872 | NP_001239212.1 | |
NM_001252287.1 | 21 | 2915 | NP_001239216.1 | |
NM_001252288.1 | 21 | 2887 | NP_001239217.1 | |
NM_010956.4 | 21 | 2839 | NP_035086.2 | |
AK147289.1 | 21 | 2840 | BAE27824.1 | |
AK150009.1 | 21 | 2805 | BAE29234.1 | |
AK169286.1 | 21 | 2831 | BAE41044.1 | |
AK190496.1 | 2 | 162 | ||
AK220536.1 | 22 | 2846 | BAD90530.1 | |
BC013670.1 | 21 | 2773 | AAH13670.1 | |
BC025040.1 | 6 | 639 | AAH25040.1 | |
BC029143.1 | 6 | 649 | AAH29143.1 | |
BC031165.1 | 6 | 618 | AAH31165.1 | |
BC049104.1 | 21 | 2840 | AAH49104.1 | |
BC057354.1 | 22 | 2800 | AAH57354.1 | |
BC061232.1 | 6 | 693 | AAH61232.1 | |
U02971.1 | 3 | 232 | AAC52130.1 |
More Information
Set Membership: |
Intragenic Exonic |