Assay Details
Target Gene Details
Entrez Gene ID: | 18293 |
Gene Name: | oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide) |
Gene Aliases: |
2210403E04Rik, 2210412K19Rik, AA409584, d1401, mKIAA4192 |
Location: |
Chr.11:6291593-6359094 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ogdh | NM_001252282.1 | NP_001239211.1 | ||
NM_001252283.1 | NP_001239212.1 | |||
NM_001252287.1 | NP_001239216.1 | |||
NM_001252288.1 | NP_001239217.1 | |||
NM_010956.4 | NP_035086.2 | |||
AK147289.1 | BAE27824.1 | |||
AK150009.1 | BAE29234.1 | |||
AK169286.1 | BAE41044.1 | |||
AK201981.1 | ||||
AK204207.1 | ||||
AK220536.1 | BAD90530.1 | |||
BC013670.1 | AAH13670.1 | |||
BC025040.1 | AAH25040.1 | |||
BC029143.1 | AAH29143.1 | |||
BC031165.1 | AAH31165.1 | |||
BC049104.1 | AAH49104.1 | |||
BC057354.1 | AAH57354.1 | |||
BC061232.1 | AAH61232.1 |
More Information
Set Membership: |
Intragenic Non-exonic |