Assay Details
Target Gene Details
Entrez Gene ID: | 268448 |
Gene Name: | PHD finger protein 12 |
Gene Aliases: |
2410142K10Rik, F630045O13, PF1, mKIAA1523 |
Location: |
Chr.11:77982754-78030539 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phf12 | NM_174852.3 | 7 | 1501 | NP_777277.2 |
AK089226.1 | 7 | 1565 | ||
AK129383.1 | 4 | 648 | BAC98193.2 | |
AK141796.1 | 7 | 1405 | BAE24837.1 | |
AK155354.1 | 7 | 1542 | BAE33212.1 | |
AK170155.1 | 7 | 1589 | BAE41602.1 | |
AK170187.1 | 6 | 796 | BAE41622.1 | |
BC043080.1 | 7 | 1104 | AAH43080.1 |
More Information
Set Membership: |
Intragenic Exonic |