Assay Details
Target Gene Details
Entrez Gene ID: | 217124 |
Gene Name: | protein phosphatase 1, regulatory subunit 9B |
Gene Aliases: |
SPL, Spn |
Location: |
Chr.11:94991212-95006898 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r9b | NM_172261.3 | NP_758465.2 | ||
AY508450.1 | AAR91608.1 | |||
BC029672.1 | 1 | 460 | AAH29672.1 |