Assay Details
Target Gene Details
Entrez Gene ID: | 69338 |
Gene Name: | RIKEN cDNA 1700003D09 gene |
Gene Aliases: |
- |
Location: |
Chr.11:98350718-98358283 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700003D09Rik | NR_045477.1 | 3 | 889 | |
AK005627.1 | 3 | 889 |
Target Gene Details
Entrez Gene ID: | 19049 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 1B |
Gene Aliases: |
AU040756, DARPP-32, Darpp32 |
Location: |
Chr.11:98348738-98357795 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r1b | NM_144828.1 | NP_659077.1 | ||
AB221544.1 | ||||
AK141563.1 | BAE24736.1 | |||
AY640620.1 | AAT66920.1 | |||
AY640621.1 | AAT66921.1 | |||
BC011122.1 | AAH11122.1 | |||
BC026568.1 | AAH26568.1 | |||
BC031129.1 | AAH31129.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |