Assay Details
Target Gene Details
Entrez Gene ID: | 56218 |
Gene Name: | POZ (BTB) and AT hook containing zinc finger 1 |
Gene Aliases: |
8430401L15Rik, AI447519, Mazr, Patz, Zfp278 |
Location: |
Chr.11:3289131-3309083 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Patz1 | NM_001253690.1 | NP_001240619.1 | ||
NM_001253691.1 | NP_001240620.1 | |||
NM_019574.3 | NP_062520.1 | |||
NR_045108.2 | ||||
NR_045109.2 | ||||
AB029397.1 | BAA90874.1 | |||
AK018372.1 | ||||
AK166189.1 | BAE38619.1 | |||
AK181924.1 | ||||
BC026673.1 | AAH26673.1 | |||
BC043035.1 | AAH43035.1 | |||
BC055947.1 | AAH55947.1 | |||
CA327687.1 |
More Information
Set Membership: |
Intragenic Non-exonic |