Assay Details
Target Gene Details
Entrez Gene ID: | 67283 |
Gene Name: | solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19 |
Gene Aliases: |
2900089E13Rik, DNC, MUP1, TPC |
Location: |
Chr.11:115614170-115628295 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a19 | NM_001252384.1 | 3 | 525 | NP_001239313.1 |
NM_001252394.1 | 2 | 362 | NP_001239323.1 | |
NM_001252395.1 | 2 | 362 | NP_001239324.1 | |
NM_001252396.1 | NP_001239325.1 | |||
NM_026071.3 | 2 | 362 | NP_080347.1 | |
AK005710.1 | 2 | 207 | BAB24199.1 | |
AK168145.1 | 2 | 161 | ||
AK172622.1 | 1 | 164 | BAE43100.1 | |
AK179517.1 | ||||
BC018167.1 | 2 | 168 | AAH18167.1 | |
BC046767.1 | 2 | 212 | AAH46767.1 | |
BI736130.1 | 2 | 174 | ||
BY276424.1 | 2 | 364 | ||
CF730205.1 | ||||
CX241396.1 | 3 | 331 |
More Information
Set Membership: |
Intragenic Exonic |