Assay Details
Target Gene Details
Entrez Gene ID: | 216578 |
Gene Name: | poly(A) polymerase gamma |
Gene Aliases: |
9630006B20Rik |
Location: |
Chr.11:23860145-23895296 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Papolg | NM_172555.2 | 19 | 2128 | NP_766143.2 |
AK035810.1 | 19 | 2111 | BAC29194.1 | |
BC019635.1 | 4 | 481 | AAH19635.1 | |
BC023030.1 | 4 | 465 | AAH23030.1 | |
BC059271.1 | 19 | 2122 | AAH59271.1 | |
DQ103505.1 | 19 | 1934 | AAZ80292.1 |
More Information
Set Membership: |
Intragenic Exonic |