Assay Details
Target Gene Details
Entrez Gene ID: | 74136 |
Gene Name: | SEC14-like 1 (S. cerevisiae) |
Gene Aliases: |
1200017E04Rik, 2810012L19Rik, Naa-35 |
Location: |
Chr.11:117115172-117159268 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sec14l1 | NM_001166506.1 | NP_001159978.1 | ||
NM_001166507.1 | NP_001159979.1 | |||
NM_028777.3 | NP_083053.2 | |||
AK004818.1 | BAB23589.1 | |||
AK172947.1 | BAD32225.1 | |||
AK195036.1 | ||||
AK200242.1 | ||||
BC005766.1 | AAH05766.1 |