Assay Details
Target Gene Details
Entrez Gene ID: | 67338 |
Gene Name: | ring finger and FYVE like domain containing protein |
Gene Aliases: |
1700051E09Rik, 4930516L10Rik, BG080975, Carp2 |
Location: |
Chr.11:82803819-82871239 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rffl | NM_001007465.3 | NP_001007466.1 | ||
NM_001164569.1 | NP_001158041.1 | |||
NM_001164570.1 | NP_001158042.1 | |||
NM_001164571.1 | NP_001158043.1 | |||
AK128983.1 | BAC87665.1 | |||
AK150213.1 | BAE29383.1 | |||
AK150541.1 | BAE29644.1 | |||
AK159442.1 | ||||
AK169393.1 | BAE41140.1 | |||
AK169988.1 | ||||
CJ097615.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |