Assay Details
Target Gene Details
Entrez Gene ID: | 18626 |
Gene Name: | period circadian clock 1 |
Gene Aliases: |
Per, m-rigui, mPer1 |
Location: |
Chr.11:69095140-69109965 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Per1 | NM_001159367.1 | NP_001152839.1 | ||
NM_011065.4 | NP_035195.2 | |||
AB002108.1 | BAA22634.1 | |||
AF022992.1 | AAC53355.1 | |||
AK081813.1 | BAC38339.1 | |||
AK148202.1 | BAE28412.1 | |||
AK154900.1 | BAE32911.1 | |||
AK172958.1 | BAD32236.1 | |||
AK182563.1 | ||||
BC039768.1 | AAH39768.1 | |||
BC091645.1 | AAH91645.1 |
More Information
Set Membership: |
Intragenic Non-exonic |