Assay Details
Target Gene Details
Entrez Gene ID: | 18626 |
Gene Name: | period circadian clock 1 |
Gene Aliases: |
Per, m-rigui, mPer1 |
Location: |
Chr.11:69095140-69109965 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Per1 | NM_001159367.1 | 16 | 2135 | NP_001152839.1 |
NM_011065.4 | 16 | 2150 | NP_035195.2 | |
AB002108.1 | 15 | 1964 | BAA22634.1 | |
AF022992.1 | 16 | 2189 | AAC53355.1 | |
AK081813.1 | 16 | 2148 | BAC38339.1 | |
AK148202.1 | 16 | 2142 | BAE28412.1 | |
AK154900.1 | 16 | 2131 | BAE32911.1 | |
AK172958.1 | 16 | 2135 | BAD32236.1 | |
BC039768.1 | 15 | 2035 | AAH39768.1 | |
BC091645.1 | 16 | 2137 | AAH91645.1 |
More Information
Set Membership: |
Intragenic Exonic |