Assay Details
Target Gene Details
Entrez Gene ID: | 52892 |
Gene Name: | SCO cytochrome oxidase deficient homolog 1 (yeast) |
Gene Aliases: |
2610001C07Rik, D11Bwg1310e |
Location: |
Chr.11:67052670-67067444 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sco1 | NM_001040026.1 | 4 | 572 | NP_001035115.1 |
AK011251.1 | 4 | 588 | ||
AK017794.1 | 4 | 573 | ||
AK139512.1 | 4 | 543 | BAE24044.1 | |
AK167305.1 | 4 | 437 | BAE39409.1 | |
BC139008.1 | 4 | 567 | ||
BC139009.1 | 4 | 567 |
More Information
Set Membership: |
Intragenic Exonic |