Assay Details
Target Gene Details
Entrez Gene ID: | 70310 |
Gene Name: | phospholipid scramblase 3 |
Gene Aliases: |
2210403O21Rik, 2610037N06Rik, ESTM3, X83310 |
Location: |
Chr.11:69846251-69852058 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plscr3 | NM_001168497.1 | NP_001161969.1 | ||
NM_023564.4 | NP_076053.1 | |||
NR_033133.1 | ||||
AF159850.1 | AAF89526.1 | |||
AK011716.1 | BAB27797.1 | |||
AK029154.1 | BAC26329.1 | |||
AK075841.1 | BAC35999.1 | |||
AK166288.1 | BAE38684.1 | |||
AK172635.1 | BAE43107.1 | |||
AK217404.1 | ||||
BC020143.1 | AAH20143.1 |
More Information
Set Membership: |
Intragenic Non-exonic |