Assay Details
Target Gene Details
Entrez Gene ID: | 83796 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2 |
Gene Aliases: |
AW322457, Baf60b |
Location: |
Chr.11:106263171-106272972 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smarcd2 | NM_001130187.1 | NP_001123659.1 | ||
NM_031878.2 | NP_114084.2 | |||
AK159259.1 | BAE34940.1 | |||
AK166123.1 | BAE38583.1 | |||
BC005732.1 | AAH05732.1 |
More Information
Set Membership: |
Intragenic Non-exonic |