Assay Details
Target Gene Details
Entrez Gene ID: | 68662 |
Gene Name: | secretoglobin, family 3A, member 1 |
Gene Aliases: |
HIN-1, LuLeu2, Pnsp2, UGRP2, Utgrp2 |
Location: |
Chr.11:49663595-49665118 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Scgb3a1 | NM_054037.2 | NP_473378.1 | ||
NM_170727.2 | NP_733923.1 | |||
AF313456.1 | AAL26216.1 | |||
AF313457.1 | AAN62328.1 | |||
AK003988.1 | ||||
BC086910.1 | AAH86910.1 |