Assay Details
Target Gene Details
Entrez Gene ID: | 76293 |
Gene Name: | microfibrillar-associated protein 4 |
Gene Aliases: |
1110007F23Rik |
Location: |
Chr.11:61485378-61488706 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mfap4 | NM_029568.2 | NP_083844.1 | ||
AK003537.1 | BAB22844.1 | |||
AK075659.1 | ||||
BC022666.1 | AAH22666.1 |
More Information
Set Membership: |
Intragenic Non-exonic |