Assay Details
Target Gene Details
Entrez Gene ID: | 23939 |
Gene Name: | mitogen-activated protein kinase 7 |
Gene Aliases: |
BMK-1, BMK1, ERK-5, ERK5, Erk5-T, PRKM7 |
Location: |
Chr.11:61488812-61494266 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mapk7 | NM_001291033.1 | 1 | 116 | NP_001277962.1 |
NM_001291034.1 | 1 | 116 | NP_001277963.1 | |
NM_001291035.1 | 1 | 116 | NP_001277964.1 | |
NM_001291036.1 | 1 | 116 | NP_001277965.1 | |
NM_001291037.1 | 1 | 116 | NP_001277966.1 | |
NM_011841.2 | 1 | 116 | NP_035971.1 | |
AB019373.1 | 1 | 66 | BAA82039.1 | |
AF126161.1 | AAD39396.1 | |||
AK082552.1 | 1 | 49 | ||
AK148119.1 | BAE28357.1 | |||
AK155187.1 | BAE33103.1 | |||
AY534740.1 | 1 | 50 | AAS38576.1 | |
BC100398.1 | 1 | 58 | AAI00399.1 | |
BY207876.1 | 1 | 146 | ||
CA320276.1 |
More Information
Set Membership: |
Intragenic Exonic |