Assay Details
Target Gene Details
Entrez Gene ID: | 216850 |
Gene Name: | KDM1 lysine (K)-specific demethylase 6B |
Gene Aliases: |
1700064E03Rik, BC038313, Jmjd3, mKIAA0346 |
Location: |
Chr.11:69398508-69418148 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kdm6b | NM_001017426.1 | 18 | 4625 | NP_001017426.1 |
AK129121.1 | 8 | 1417 | BAC97931.1 | |
AK136019.1 | 8 | 806 | BAE22775.1 | |
BC075632.1 | 19 | 4801 | AAH75632.1 |
More Information
Set Membership: |
Intragenic Exonic |