Assay Details
Target Gene Details
Entrez Gene ID: | 16407 |
Gene Name: | integrin alpha E, epithelial-associated |
Gene Aliases: |
A530055J10, CD103, aM290, alpha-E1, alpha-M290 |
Location: |
Chr.11:73090595-73147449 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgae | NM_008399.2 | 6 | 534 | NP_032425.2 |
NM_172944.2 | 6 | 534 | NP_766532.1 | |
AK040983.1 | 6 | 535 | BAC30769.1 | |
AK154021.1 | 6 | 532 | BAE32321.1 | |
BC150690.1 | 6 | 534 | ||
DQ386608.1 | 6 | 534 | ||
U12236.1 | 6 | 558 | AAC52142.1 |
More Information
Set Membership: |
Intragenic Exonic |