Assay Details
Target Gene Details
Entrez Gene ID: | 70357 |
Gene Name: | Kv channel-interacting protein 1 |
Gene Aliases: |
KCHIP1, Kchip1.2 |
Location: |
Chr.11:33629339-33993193 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnip1 | NM_001190885.1 | NP_001177814.1 | ||
NM_001190886.1 | NP_001177815.1 | |||
NM_001290690.1 | NP_001277619.1 | |||
NM_027398.3 | NP_081674.2 | |||
AB075041.1 | BAB78543.1 | |||
AJ278534.1 | CAC82025.1 | |||
AK014313.1 | ||||
AK045948.1 | BAC32543.1 | |||
AK046398.1 | BAC32705.1 | |||
AK048178.1 | ||||
AK081845.1 | BAC38347.1 | |||
AK082267.1 | ||||
AY050525.1 | AAL12488.1 | |||
AY050526.1 | AAL12489.1 | |||
AY171234.1 | AAN77492.1 | |||
AY647242.1 | AAT68468.1 | |||
BC034241.1 | AAH34241.1 | |||
BU702946.1 | ||||
DQ148493.1 | AAZ77810.1 | |||
DQ148494.1 | AAZ77811.1 | |||
DQ148495.1 | AAZ77812.1 |