Assay Details
Target Gene Details
Entrez Gene ID: | 238023 |
Gene Name: | hexosaminidase (glycosyl hydrolase family 20, catalytic domain) containing |
Gene Aliases: |
BC069960 |
Location: |
Chr.11:121204433-121222656 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hexdc | NM_001001333.2 | 8 | 1222 | NP_001001333.2 |
NM_001146073.1 | 7 | 808 | NP_001139545.1 | |
AK154237.1 | 8 | 1196 | BAE32455.1 | |
BC069960.1 | 7 | 760 | AAH69960.1 | |
FM204885.1 | 7 | 759 | ||
FM204886.1 | 7 | 759 |