Assay Details
Target Gene Details
Entrez Gene ID: | 66213 |
Gene Name: | mediator complex subunit 7 |
Gene Aliases: |
1110063B05Rik, Crsp33, Crsp9 |
Location: |
Chr.11:46436933-46442721 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Med7 | NM_001104530.1 | NP_001098000.1 | ||
NM_001104557.1 | NP_001098027.1 | |||
BY135693.1 | ||||
CK030901.1 |
More Information
Set Membership: |
Intragenic Non-exonic |