Assay Details
Target Gene Details
Entrez Gene ID: | 338367 |
Gene Name: | myosin ID |
Gene Aliases: |
9930104H07Rik, AW544947, D11Ertd9e, myosin-1d |
Location: |
Chr.11:80482126-80780055 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myo1d | NM_177390.3 | 16 | 2291 | NP_796364.2 |
AK049374.1 | 16 | 2292 | BAC33719.1 | |
AK148112.1 | 3 | 410 | BAE28351.1 | |
AK190949.1 | ||||
BC034071.1 | 2 | 271 | AAH34071.1 | |
BC150765.1 | 16 | 2157 |
More Information
Set Membership: |
Intragenic Exonic |