Assay Details
Target Gene Details
Entrez Gene ID: | 23989 |
Gene Name: | mediator complex subunit 24 |
Gene Aliases: |
911GSE, AU040102, AW547152, D11Ertd307e, DRIP100, Gse2, Pparb2, Pparbp2, R75526, Thrap4, Trap100 |
Location: |
Chr.11:98704591-98729475 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Med24 | NM_011869.2 | 2 | 162 | NP_035999.2 |
AF126543.1 | 2 | 176 | AAD42776.1 | |
AK020269.1 | 2 | 137 | BAB32051.1 | |
AK154618.1 | 2 | 147 | BAE32717.1 | |
AK158385.1 | 2 | 156 | ||
AK192144.1 | 1 | 109 | ||
AK193218.1 | 1 | 109 | ||
BC005409.1 | 2 | 184 | AAH05409.1 | |
BY034665.1 | 2 | 163 | ||
CX566267.1 | 2 | 135 |
More Information
Set Membership: |
Intragenic Exonic |