Assay Details
Target Gene Details
Entrez Gene ID: | 14534 |
Gene Name: | K(lysine) acetyltransferase 2A |
Gene Aliases: |
1110051E14Rik, AW212720, Gcn5, Gcn5l2, mmGCN5 |
Location: |
Chr.11:100704746-100712467 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat2a | NM_001038010.2 | 12 | 1860 | NP_001033099.1 |
NM_020004.5 | 12 | 1863 | NP_064388.2 | |
AF254441.1 | 12 | 1800 | AAF70497.1 | |
AK137358.1 | 12 | 1840 | BAE23321.1 | |
AK145205.1 | 10 | 1860 | BAE26296.1 | |
AK147042.1 | 12 | 1808 | ||
AK150225.1 | 12 | 1841 | BAE29392.1 | |
AK154866.1 | 12 | 1851 | ||
AK155347.1 | 12 | 2037 | BAE33207.1 | |
AK155791.1 | 12 | 1862 | ||
AK155842.1 | 12 | 1861 | BAE33458.1 | |
AK158079.1 | 12 | 1851 | BAE34351.1 | |
AK186107.1 | 2 | 126 | ||
BC063752.1 | 12 | 1845 | AAH63752.1 |