Assay Details
Target Gene Details
Entrez Gene ID: | 16399 |
Gene Name: | integrin alpha 2b |
Gene Aliases: |
AI172977, CD41, CD41B, GpIIb, alphaIIb |
Location: |
Chr.11:102453297-102469883 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itga2b | NM_010575.2 | 30 | 3293 | NP_034705.2 |
AF170316.1 | 30 | 3154 | AAD56216.1 | |
AK140749.1 | 25 | 2945 | ||
AK142289.1 | 30 | 3155 | BAE25013.1 | |
AK154619.1 | 30 | 3294 | BAE32718.1 | |
AK156872.1 | 29 | 3474 | ||
AK157012.1 | 29 | 3478 | ||
AK158443.1 | 30 | 3145 | ||
BC120493.1 | 30 | 3143 | ||
BC137570.1 | 30 | 3143 |
More Information
Set Membership: |
Intragenic Exonic |