Assay Details
Target Gene Details
Entrez Gene ID: | 15184 |
Gene Name: | histone deacetylase 5 |
Gene Aliases: |
AI426555, Hdac4, mHDA1, mKIAA0600 |
Location: |
Chr.11:102195747-102230186 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac5 | NM_001077696.1 | 8 | 1188 | NP_001071164.1 |
NM_001284248.1 | 10 | 1700 | NP_001271177.1 | |
NM_001284249.1 | 9 | 1297 | NP_001271178.1 | |
NM_001284250.1 | 8 | 1185 | NP_001271179.1 | |
NM_010412.3 | 8 | 1185 | NP_034542.3 | |
AF006602.2 | 7 | 901 | AAD09834.2 | |
AF207748.1 | 7 | 820 | AAF31418.1 | |
AK122325.2 | 3 | 224 | BAC65607.2 | |
AK139202.1 | 9 | 1271 | ||
AK145972.1 | 8 | 1151 | ||
AK146483.1 | 8 | 1124 | BAE27204.1 | |
AK155335.1 | 8 | 1159 | BAE33201.1 | |
AK156008.1 | 5 | 1436 | ||
AK157159.1 | 5 | 1436 | ||
AK196092.1 | 1 | 49 | ||
AK200052.1 | 1 | 49 | ||
BC060609.1 | 7 | 979 | AAH60609.1 |
More Information
Set Membership: |
Intragenic Exonic |