Assay Details
Target Gene Details
Entrez Gene ID: | 15184 |
Gene Name: | histone deacetylase 5 |
Gene Aliases: |
AI426555, Hdac4, mHDA1, mKIAA0600 |
Location: |
Chr.11:102195747-102230186 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac5 | NM_001077696.1 | 11 | 1579 | NP_001071164.1 |
NM_001284248.1 | 13 | 2091 | NP_001271177.1 | |
NM_001284249.1 | 12 | 1688 | NP_001271178.1 | |
NM_001284250.1 | 11 | 1576 | NP_001271179.1 | |
NM_010412.3 | 11 | 1576 | NP_034542.3 | |
AF006602.2 | 10 | 1292 | AAD09834.2 | |
AF207748.1 | 10 | 1211 | AAF31418.1 | |
AK122325.2 | 6 | 615 | BAC65607.2 | |
AK139202.1 | 12 | 1662 | ||
AK145972.1 | 11 | 1542 | ||
AK146483.1 | 11 | 1515 | BAE27204.1 | |
AK155335.1 | 11 | 1550 | BAE33201.1 | |
AK156008.1 | 5 | 2413 | ||
AK157159.1 | 5 | 2413 | ||
AK191268.1 | 2 | 142 | ||
AK205148.1 | ||||
BC060609.1 | 10 | 1370 | AAH60609.1 |
More Information
Set Membership: |
Intragenic Exonic |