Assay Details
Target Gene Details
Entrez Gene ID: | 14783 |
Gene Name: | growth factor receptor bound protein 10 |
Gene Aliases: |
5730571D09Rik, AI325020, Meg1, mKIAA0207 |
Location: |
Chr.11:11930499-12037422 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grb10 | NM_001177629.1 | 4 | 444 | NP_001171100.1 |
NM_010345.4 | 4 | 604 | NP_034475.2 | |
AF022072.1 | 4 | 578 | AAB72103.1 | |
AK012646.1 | 4 | 584 | ||
AK030727.1 | 4 | 539 | BAC27100.1 | |
AK032927.1 | 4 | 585 | BAC28088.1 | |
AK163841.1 | 4 | 585 | BAE37514.1 | |
BC016111.1 | 4 | 444 | AAH16111.1 | |
BC053842.1 | 4 | 571 | AAH53842.1 |
More Information
Set Membership: |
Intragenic Exonic |