Assay Details
Target Gene Details
Entrez Gene ID: | 20185 |
Gene Name: | nuclear receptor co-repressor 1 |
Gene Aliases: |
5730405M06Rik, A230020K14Rik, N-CoR, RIP13, Rxrip13, mKIAA1047 |
Location: |
Chr.11:62315643-62457332 on Build GRCm38 |
Assay Gene Location: | Within Exon 40 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncor1 | NM_001252313.1 | 38 | 6177 | NP_001239242.1 |
NM_011308.3 | 38 | 5976 | NP_035438.3 | |
AB093281.1 | 20 | 3917 | BAC41465.1 | |
AK020951.1 | 1 | 551 | ||
AK137632.1 | 12 | 2295 | BAE23439.1 | |
BC086657.1 | 38 | 5942 | AAH86657.1 | |
L78294.1 | 17 | 2984 | AAL40135.1 | |
U22016.1 | 4 | 731 | AAC52168.1 | |
U35312.1 | 37 | 6220 | AAB17125.1 |
More Information
Set Membership: |
Intragenic Exonic |