Assay Details
Target Gene Details
Entrez Gene ID: | 14778 |
Gene Name: | glutathione peroxidase 3 |
Gene Aliases: |
AA960521, EGPx, GPx, GSHPx-3, GSHPx-P |
Location: |
Chr.11:54902854-54910382 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpx3 | NM_008161.3 | 2 | 238 | NP_032187.2 |
AK002219.1 | 2 | 167 | BAC55243.1 | |
AK002262.1 | 2 | 167 | ||
AK004942.1 | 2 | 167 | BAC55250.1 | |
AK146760.1 | 2 | 167 | BAE27413.1 | |
BC003339.1 | 2 | 150 | AAH03339.1 | |
BC037027.1 | 2 | 185 | AAH37027.1 | |
BC049235.1 | 2 | 154 | AAH49235.1 | |
BC061950.1 | 2 | 142 | AAH61950.1 | |
BY061328.1 | 2 | 239 | ||
BY100863.1 | 2 | 240 | ||
BY339672.1 | 2 | 240 | ||
CB601893.1 | 2 | 170 | ||
U13705.1 | 2 | 142 | AAA62283.2 |
More Information
Set Membership: |
Intragenic Exonic |