Assay Details
Target Gene Details
Entrez Gene ID: | 14778 |
Gene Name: | glutathione peroxidase 3 |
Gene Aliases: |
AA960521, EGPx, GPx, GSHPx-3, GSHPx-P |
Location: |
Chr.11:54902854-54910382 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpx3 | NM_008161.3 | NP_032187.2 | ||
BY061328.1 | ||||
BY100863.1 | ||||
BY339672.1 |
More Information
Set Membership: |
Intragenic Non-exonic |