Assay Details
Target Gene Details
Entrez Gene ID: | 17428 |
Gene Name: | max binding protein |
Gene Aliases: |
Rox, bHLHd3 |
Location: |
Chr.11:74830796-74845725 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mnt | NM_010813.3 | 5 | 1271 | NP_034943.3 |
AK139743.1 | 4 | 293 | ||
AK147933.1 | 5 | 1255 | BAE28239.1 | |
BC054534.1 | 5 | 1197 | AAH54534.1 | |
U77356.1 | 5 | 1271 | AAB38687.1 | |
Y07609.1 | 5 | 1217 | CAA68878.1 |
More Information
Set Membership: |
Intragenic Exonic |