Assay Details
Target Gene Details
Entrez Gene ID: | 69674 |
Gene Name: | MIF4G domain containing |
Gene Aliases: |
1110014L05Rik, 2310075G12Rik |
Location: |
Chr.11:115607918-115612973 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mif4gd | NM_001243584.1 | NP_001230513.1 | ||
NM_001243586.1 | NP_001230515.1 | |||
NM_001243587.1 | NP_001230516.1 | |||
NM_027162.4 | NP_081438.4 | |||
AK010175.1 | ||||
AK150749.1 | BAE29819.1 | |||
BC019515.1 | ||||
BC026740.1 | AAH26740.1 | |||
BC026745.1 | ||||
BC055812.1 | AAH55812.1 | |||
CX210298.1 |
More Information
Set Membership: |
Intragenic Non-exonic |