Assay Details
Target Gene Details
Entrez Gene ID: | 192897 |
Gene Name: | integrin beta 4 |
Gene Aliases: |
AA407042, C230078O20, CD104 |
Location: |
Chr.11:115974725-116008412 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb4 | NM_001005608.2 | 24 | 3148 | NP_001005608.2 |
NM_133663.2 | 24 | 3148 | NP_598424.2 | |
AK048888.1 | 24 | 3033 | ||
AK144111.1 | 1 | 144 | BAE25705.1 | |
AK145317.1 | 1 | 224 | BAE26363.1 | |
AK147424.1 | 24 | 3146 | ||
BC006632.1 | 4 | 194 | AAH06632.1 | |
BC059192.1 | 20 | 2343 | AAH59192.1 | |
L04678.1 | 25 | 2992 |
More Information
Set Membership: |
Intragenic Exonic |