Assay Details
Target Gene Details
Entrez Gene ID: | 21428 |
Gene Name: | MAX-like protein X |
Gene Aliases: |
Tcfl4, bHLHd13 |
Location: |
Chr.11:101086709-101092456 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mlx | NM_001159384.1 | NP_001152856.1 | ||
NM_001159385.1 | NP_001152857.1 | |||
NM_011550.3 | NP_035680.3 | |||
AF213670.1 | AAG40149.1 | |||
AF213671.1 | AAG40150.1 | |||
AF213672.1 | AAG40151.1 | |||
AK011478.1 | ||||
AK142605.1 | BAE25128.1 | |||
BC031380.1 | AAH31380.1 | |||
U43548.1 | AAB51367.1 | |||
U43607.1 | AAB51368.1 |
More Information
Set Membership: |
Intragenic Non-exonic |